Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTR4
Entrez Gene ID
317708
Gene Name
5-hydroxytryptamine receptor 4
Gene Aliases
-
Chromosome Location
Chr.7: 59872178 - 60080245 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040485.1 NP_001035575.1 4 - 5 449 87
GenBank mRNA BT020865.1 - 4 - 5 449 87

View Full Product Details