Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LGI1
Entrez Gene ID
617080
Gene Name
leucine rich glioma inactivated 1
Gene Aliases
-
Chromosome Location
Chr.26: 15237501 - 15280182 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046591.2 NP_001040056.2 2 - 3 568 146
GenBank mRNA BT020834.1 - 2 - 3 568 146

View Full Product Details