Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AKR1A1
Entrez Gene ID
618607
Gene Name
aldo-keto reductase family 1 member A1
Gene Aliases
-
Chromosome Location
Chr.3: 100505185 - 100516929 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076513.2 NP_001069981.1 7 - 8 941 62
GenBank mRNA BC102166.1 - 7 - 8 942 62

View Full Product Details