Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACR
Entrez Gene ID
280711
Gene Name
acrosin
Gene Aliases
-
Chromosome Location
Chr.5: 120013292 - 120020813 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173886.2 NP_776311.2 3 - 4 576 89
GenBank mRNA EV607234.1 - 3 - 4 583 89

View Full Product Details