Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LDHB
Entrez Gene ID
281275
Gene Name
lactate dehydrogenase B
Gene Aliases
-
Chromosome Location
Chr.5: 88544523 - 88562770 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316338.1 NP_001303267.1 4 - 5 567 114
NM_174100.2 NP_776525.2 4 - 5 567 114
GenBank mRNA AB099066.1 - 1 - 2 77 115
AJ401268.1 - 4 - 5 531 114
BC102217.1 - 4 - 5 498 114
BC142006.1 - 4 - 5 568 114
BC151427.1 - 4 - 5 498 114
BT021009.1 - 4 - 5 535 114
DT720873.1 - 4 - 5 508 114

View Full Product Details