Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGDIA
Entrez Gene ID
338054
Gene Name
Rho GDP dissociation inhibitor alpha
Gene Aliases
-
Chromosome Location
Chr.19: 51023538 - 51028507 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176650.3 NP_788823.1 2 - 3 262 108
GenBank mRNA BC103465.1 - 2 - 3 262 108
BT021511.1 - 2 - 3 243 108
X52689.1 - - 232 108

View Full Product Details