Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGDIA
Entrez Gene ID
338054
Gene Name
Rho GDP dissociation inhibitor alpha
Gene Aliases
-
Chromosome Location
Chr.19: 51023538 - 51028507 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176650.3 NP_788823.1 3 - 4 355 105
GenBank mRNA BC103465.1 - 3 - 4 355 105
BT021511.1 - 3 - 4 336 105
X52689.1 - - 325 105

View Full Product Details