Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL18
Entrez Gene ID
509163
Gene Name
ribosomal protein L18
Gene Aliases
-
Chromosome Location
Chr.18: 55260179 - 55263944 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015556.2 NP_001015556.1 2 - 2 511 66
GenBank mRNA BC104507.1 - 2 - 2 511 66
BT021047.1 - 6 - 7 510 66
DQ403039.1 - 5 - 6 419 66

View Full Product Details