Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL27
Entrez Gene ID
404137
Gene Name
ribosomal protein L27
Gene Aliases
-
Chromosome Location
Chr.19: 43026105 - 43028407 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034051.2 NP_001029223.1 4 - 5 407 76
GenBank mRNA AB098983.1 - 4 - 5 389 76
BC102313.1 - 4 - 5 409 76

View Full Product Details