Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR2C
Entrez Gene ID
504452
Gene Name
RNA polymerase II subunit C
Gene Aliases
-
Chromosome Location
Chr.18: 25458202 - 25466511 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034212.2 NP_001029384.1 4 - 5 285 111
GenBank mRNA BC102301.1 - 4 - 5 287 111
BT030680.1 - 4 - 5 281 111

View Full Product Details