Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NSA2
Entrez Gene ID
522309
Gene Name
NSA2 ribosome biogenesis homolog (S. cerevisiae)
Gene Aliases
TINP1
Chromosome Location
Chr.20: 6742910 - 6750047 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034546.2 NP_001029718.1 1 - 2 118 90
GenBank mRNA BC104563.1 - 1 - 2 119 90

View Full Product Details