Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMB3
Entrez Gene ID
533874
Gene Name
proteasome 20S subunit beta 3
Gene Aliases
-
Chromosome Location
Chr.19: 39344060 - 39353531 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034596.2 NP_001029768.1 4 - 5 562 79
GenBank mRNA BC102554.1 - 4 - 5 563 79
BC151435.1 - 4 - 5 562 79

View Full Product Details