Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECM2
Entrez Gene ID
533916
Gene Name
extracellular matrix protein 2
Gene Aliases
-
Chromosome Location
Chr.8: 84105569 - 84148313 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034597.1 NP_001029769.1 9 - 10 1996 71
GenBank mRNA BC105232.1 - 9 - 10 1996 71

View Full Product Details