Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB28
Entrez Gene ID
536021
Gene Name
RAB28, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.6: 108599626 - 108697605 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034634.1 NP_001029806.1 3 - 4 474 121
GenBank mRNA BC104592.1 - 3 - 4 474 121

View Full Product Details