Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZD3
Entrez Gene ID
539171
Gene Name
PDZ domain containing 3
Gene Aliases
-
Chromosome Location
Chr.15: 29769699 - 29775955 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034664.1 NP_001029836.1 1 - 2 24 65
GenBank mRNA BC105272.1 - 1 - 2 24 65

View Full Product Details