Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSD3B7
Entrez Gene ID
540229
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
Gene Aliases
-
Chromosome Location
Chr.25: 27119053 - 27122803 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034696.1 NP_001029868.1 3 - 4 400 67
GenBank mRNA BC105259.1 - 3 - 4 400 67
BT030700.1 - 3 - 4 401 67

View Full Product Details