Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINC1
Entrez Gene ID
540261
Gene Name
serpin family C member 1
Gene Aliases
AT3
Chromosome Location
Chr.16: 55267228 - 55277841 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034698.2 NP_001029870.1 6 - 7 1288 77
GenBank mRNA BC102747.1 - 6 - 7 1289 77

View Full Product Details