Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACYBP
Entrez Gene ID
618428
Gene Name
calcyclin binding protein
Gene Aliases
-
Chromosome Location
Chr.16: 56231860 - 56242613 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034809.2 NP_001029981.1 1 - 2 124 117
GenBank mRNA BC102091.1 - 1 - 2 126 117

View Full Product Details