Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DERL2
Entrez Gene ID
508465
Gene Name
derlin 2
Gene Aliases
-
Chromosome Location
Chr.19: 26073470 - 26082010 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035059.2 NP_001030231.2 6 - 7 642 106
GenBank mRNA BC102071.1 - 5 - 6 550 106
EH210391.1 - 6 - 7 642 106
EV741886.1 - 1 - 2 533 106

View Full Product Details