Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDCA3
Entrez Gene ID
614434
Gene Name
cell division cycle associated 3
Gene Aliases
-
Chromosome Location
Chr.5: 103598733 - 103600977 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035449.2 NP_001030526.1 2 - 3 236 76
GenBank mRNA BC102247.1 - 2 - 3 237 76

View Full Product Details