Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMS
Entrez Gene ID
615950
Gene Name
spermine synthase
Gene Aliases
-
Chromosome Location
Chr.X: 121317041 - 121367225 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035471.1 NP_001030548.1 8 - 9 975 121
GenBank mRNA BC103012.1 - 8 - 9 975 121
BC151416.1 - 8 - 9 901 121
BT029883.1 - 8 - 9 928 121

View Full Product Details