Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1D
Entrez Gene ID
506667
Gene Name
C1D nuclear receptor corepressor
Gene Aliases
-
Chromosome Location
Chr.11: 66490747 - 66513682 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037448.2 NP_001032525.1 1 - 2 53 119
GenBank mRNA BC108147.1 - 1 - 2 54 119

View Full Product Details