Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MON1B
Entrez Gene ID
510379
Gene Name
MON1 homolog B, secretory trafficking associated
Gene Aliases
-
Chromosome Location
Chr.18: 4411467 - 4423090 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037454.2 NP_001032531.1 1 - 2 347 66
GenBank mRNA BC108085.1 - 1 - 2 348 66

View Full Product Details