Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUV39H2
Entrez Gene ID
536936
Gene Name
suppressor of variegation 3-9 homolog 2
Gene Aliases
-
Chromosome Location
Chr.13: 29527149 - 29551011 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037479.2 NP_001032556.1 1 - 2 55 144
GenBank mRNA BC108111.1 - 1 - 2 56 144

View Full Product Details