Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSR4
Entrez Gene ID
504438
Gene Name
signal sequence receptor subunit 4
Gene Aliases
-
Chromosome Location
Chr.X: 37011611 - 37015530 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038503.2 NP_001033592.1 4 - 5 380 79
GenBank mRNA BC109500.1 - 4 - 5 381 79

View Full Product Details