Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF280B
Entrez Gene ID
517697
Gene Name
zinc finger protein 280B
Gene Aliases
SUHW2
Chromosome Location
Chr.17: 70979561 - 70990872 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077935.2 NP_001071403.2 2 - 3 188 66
GenBank mRNA BC102316.1 - 2 - 3 158 66

View Full Product Details