Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNTN
Entrez Gene ID
768326
Gene Name
sentan, cilia apical structure protein
Gene Aliases
S100A1L, sentan
Chromosome Location
Chr.22: 37750448 - 37764990 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078138.2 NP_001071606.1 3 - 4 304 109
GenBank mRNA BC120476.1 - 3 - 4 305 109

View Full Product Details