Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC8
Entrez Gene ID
777785
Gene Name
thioredoxin domain containing 8
Gene Aliases
-
Chromosome Location
Chr.8: 99835325 - 99867778 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078160.1 NP_001071628.1 3 - 4 255 73
GenBank mRNA BC126781.1 - 3 - 4 255 73

View Full Product Details