Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RXRB
Entrez Gene ID
504943
Gene Name
retinoid X receptor beta
Gene Aliases
-
Chromosome Location
Chr.23: 7379995 - 7386713 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083640.1 NP_001077109.1 3 - 4 638 72
GenBank mRNA BC146260.1 - 3 - 4 817 72

View Full Product Details