Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SF3B5
Entrez Gene ID
574400
Gene Name
splicing factor 3b subunit 5
Gene Aliases
-
Chromosome Location
Chr.9: 81491288 - 81492016 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025350.3 NP_001020521.1 - 368 69
GenBank mRNA AY911314.1 - 1 - 2 369 69
BC111655.1 - - 369 69

View Full Product Details