Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MADCAM1
Entrez Gene ID
527275
Gene Name
mucosal vascular addressin cell adhesion molecule 1
Gene Aliases
-
Chromosome Location
Chr.7: 43158741 - 43163415 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037821.1 NP_001032910.1 2 - 3 354 138
GenBank mRNA DQ288286.1 - 2 - 3 354 138

View Full Product Details