Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RXYLT1
Entrez Gene ID
515147
Gene Name
ribitol xylosyltransferase 1
Gene Aliases
TMEM5
Chromosome Location
Chr.5: 49919034 - 49945712 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038114.2 NP_001033203.1 4 - 5 877 117
GenBank mRNA BC109876.1 - 4 - 5 879 117

View Full Product Details