Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2D1
Entrez Gene ID
535287
Gene Name
ubiquitin conjugating enzyme E2 D1
Gene Aliases
-
Chromosome Location
Chr.26: 665570 - 703300 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038167.1 NP_001033256.1 6 - 7 642 99
GenBank mRNA BC111175.1 - 6 - 7 642 99

View Full Product Details