Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIBC2
Entrez Gene ID
540723
Gene Name
RIB43A domain with coiled-coils 2
Gene Aliases
-
Chromosome Location
Chr.5: 115818287 - 115829117 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038191.2 NP_001033280.2 4 - 5 837 101
GenBank mRNA BC109543.1 - 4 - 5 838 101

View Full Product Details