Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC23
Entrez Gene ID
613700
Gene Name
leucine rich repeat containing 23
Gene Aliases
-
Chromosome Location
Chr.5: 103546870 - 103554402 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038199.2 NP_001033288.1 1 - 2 171 70
GenBank mRNA BC109993.1 - 1 - 2 172 70

View Full Product Details