Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GINM1
Entrez Gene ID
506714
Gene Name
glycoprotein integral membrane 1
Gene Aliases
C9H6orf72
Chromosome Location
Chr.9: 86800168 - 86821630 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038509.2 NP_001033598.1 4 - 5 439 147
GenBank mRNA BC110005.1 - 4 - 5 441 147

View Full Product Details