Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MED28
Entrez Gene ID
513972
Gene Name
mediator complex subunit 28
Gene Aliases
-
Chromosome Location
Chr.6: 37169216 - 37175026 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038538.1 NP_001033627.1 2 - 3 254 119
GenBank mRNA BC109886.1 - 2 - 3 254 119

View Full Product Details