Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP4
Entrez Gene ID
507503
Gene Name
THAP domain containing 4
Gene Aliases
-
Chromosome Location
Chr.3: 120665989 - 120709527 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038669.1 NP_001033758.1 3 - 4 1455 78
GenBank mRNA BC110168.1 - 3 - 4 1455 78
BC118115.1 - 2 - 3 266 78

View Full Product Details