Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HP
Entrez Gene ID
280692
Gene Name
haptoglobin
Gene Aliases
-
Chromosome Location
Chr.18: 39037405 - 39043561 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040470.2 NP_001035560.1 5 - 6 375 108
GenBank mRNA BC109668.1 - 5 - 6 376 108
KR006344.1 - 4 - 5 345 108

View Full Product Details