Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADIPOR2
Entrez Gene ID
407234
Gene Name
adiponectin receptor 2
Gene Aliases
-
Chromosome Location
Chr.5: 108184042 - 108227652 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040499.2 NP_001035589.1 6 - 7 1112 71
GenBank mRNA AY436319.1 - 2 - 3 286 71
BC110019.1 - 6 - 7 1115 71
BT025411.1 - 4 - 5 677 71

View Full Product Details