Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDCD2
Entrez Gene ID
532693
Gene Name
NudC domain containing 2
Gene Aliases
-
Chromosome Location
Chr.7: 75130669 - 75136607 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040553.2 NP_001035643.1 1 - 2 407 64
GenBank mRNA BC109920.1 - 1 - 2 409 64
BC134768.1 - 1 - 2 221 64

View Full Product Details