Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHOSPHO2
Entrez Gene ID
507308
Gene Name
phosphatase, orphan 2
Gene Aliases
-
Chromosome Location
Chr.2: 26613241 - 26618778 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045965.2 NP_001039430.1 1 - 2 208 129
GenBank mRNA BC112815.1 - 1 - 2 209 129

View Full Product Details