Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SF3B6
Entrez Gene ID
508876
Gene Name
splicing factor 3b subunit 6
Gene Aliases
SF3B14
Chromosome Location
Chr.11: 74990095 - 74998220 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046017.1 NP_001039482.1 3 - 4 517 112
GenBank mRNA BC112548.1 - 3 - 4 517 112

View Full Product Details