Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTF2E2
Entrez Gene ID
510921
Gene Name
general transcription factor IIE subunit 2
Gene Aliases
-
Chromosome Location
Chr.27: 26703211 - 26783526 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046065.2 NP_001039530.1 5 - 6 873 121
GenBank mRNA BC105362.1 - 5 - 6 874 121

View Full Product Details