Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO3
Entrez Gene ID
532870
Gene Name
LIM domain only 3
Gene Aliases
-
Chromosome Location
Chr.5: 93264607 - 93330594 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046337.2 NP_001039802.1 2 - 3 108 84
GenBank mRNA BC112712.1 - 2 - 3 109 84

View Full Product Details