Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALKBH7
Entrez Gene ID
535305
Gene Name
alkB homolog 7
Gene Aliases
-
Chromosome Location
Chr.7: 18073869 - 18075473 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046402.2 NP_001039867.1 3 - 4 522 121
GenBank mRNA BC111665.1 - 3 - 4 523 121

View Full Product Details