Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERI3
Entrez Gene ID
540516
Gene Name
ERI1 exoribonuclease family member 3
Gene Aliases
PRNPIP
Chromosome Location
Chr.3: 101701172 - 101832068 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046479.2 NP_001039944.1 3 - 4 266 83
NM_001113242.2 NP_001106713.1 5 - 6 897 83
GenBank mRNA BC110159.1 - 3 - 4 267 83
BC147967.1 - 5 - 6 899 83

View Full Product Details