Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2Q2
Entrez Gene ID
511631
Gene Name
ubiquitin conjugating enzyme E2 Q2
Gene Aliases
-
Chromosome Location
Chr.21: 31126678 - 31189226 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075530.2 NP_001068998.1 3 - 4 498 106
GenBank mRNA BC109798.1 - 3 - 4 499 106

View Full Product Details