Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB8OS
Entrez Gene ID
613795
Gene Name
zinc finger and BTB domain containing 8 opposite strand
Gene Aliases
-
Chromosome Location
Chr.2: 120986507 - 121002878 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076288.2 NP_001069756.1 2 - 3 550 65
GenBank mRNA BC110257.1 - 2 - 3 551 65

View Full Product Details