Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC4
Entrez Gene ID
618292
Gene Name
exosome component 4
Gene Aliases
RRP41
Chromosome Location
Chr.14: 755134 - 757010 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078086.2 NP_001071554.1 1 - 2 232 102
GenBank mRNA BC109819.1 - 1 - 2 233 102

View Full Product Details